Rhodium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010307-100N
RV010307-250N
RV010307-500N
RCRV010307100NEA
1559.46
CAD
RCRV010307100N
RCRV010307250N
RCRV010307500N
Rhodium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards
Cationic Standards
Rhodium Standards
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml