Platinum Standards, for ICP, VeriSpec®, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010304-500N RV010406-250N RV010406-100N RV010406-500N RV010304-250N RV010304-100N RV010304-50N
RCRV010304500NEA 4792.03 CAD
RCRV010304500N RCRV010406250N RCRV010304100N RCRV010406500N RCRV010304-50N RCRV010406100N RCRV010304250N
Platinum Standards, for ICP, VeriSpec®, RICCA Chemical Company
Standards Cationic Standards Platinum Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml