Platinum Standard, for AAS, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010041-100C RV010041-500C
RCRV010041100CEA 440.82 CAD
RCRV010041100C RCRV010041500C
Platinum Standard, for AAS, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Platinum Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml