Palladium Standards, for AAS, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010040-100C
RV010040-500C
RV010081-100C
RV010081-500C
RCRV010040100CEA
440.82
CAD
RCRV010040100C
RCRV010040500C
RCRV010081100C
RCRV010081500C
Palladium Standards, for AAS, VeriSpec®, Ricca Chemical Company
Standards
Cationic Standards
Palladium Standards
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml