Palladium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010302-250N RV010302-500N RV010347-250N RV010347-500N RV010404-100N RV010474-100N
RCRV010302100NEA 443.22 CAD
RCRV010302100N RCRV010302250N RCRV010302500N RCRV010347100N RCRV010347250N RCRV010347500N RCRV010404100N RCRV010474100N
Palladium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Palladium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml