Monomethylamine - CH₃NH₃+ Standard, for Ion Chromatography, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010927-100N RV010927-250N RV010927-500N
RCRV010927100NEA 199.08 CAD
RCRV010927100N RCRV010927250N RCRV010927500N
Monomethylamine - CH₃NH₃+ Standard, for Ion Chromatography, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Amine Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml