Monoethanolamine -H₃NC₂H₄OH+ Standard, for Ion Chromatography, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010926-100N RV010926-250N RV010926-500N
RCRV010926100NEA 180.12 CAD
RCRV010926100N RCRV010926250N RCRV010926500N
Monoethanolamine -H₃NC₂H₄OH+ Standard, for Ion Chromatography, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Amine Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml
Order Now
SPECIFICATIONS
Environmentally Preferable