Mercury Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010284-100N
RV010284-250N
RV010284-500N
RCRV010284100NEA
221.2
CAD
RCRV010284100N
RCRV010284250N
RCRV010284500N
RCRV010386100N
Mercury Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards
Cationic Standards
Mercury Standards
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml