Hafnium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010339-100N RV010339-50N RV010339-250N RV010455-100N RV010455-500N RV010339-500N RV010455-50N RV010455-250N
RCRV010339500NEA 861.57 CAD
RCRV010339500N RCRV010339-50N RCRV010455250N RCRV010339250N RCRV010339100N RCRV010455100N RCRV010455500N RCRV010455-50N
Hafnium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Hafnium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml