Gold Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010263-100N
RV010263-250N
RV010263-500N
RV010366-100N
RCRV010263-50NEA
258.77
CAD
RCRV010263-50N
RCRV010263100N
RCRV010263250N
RCRV010263500N
RCRV010366100N
Gold Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards
Cationic Standards
Gold Standards
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml