Germanium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010282-100N
RV010282-250N
RV010282-500N
RV010384-100N
RCRV010282100NEA
205.4
CAD
RCRV010282100N
RCRV010282250N
RCRV010282500N
RCRV010384100N
Germanium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards
Cationic Standards
Germanium Standards
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml