Germanium Standard, for AAS, VeriSpec®, RICCA Chemical Company
Supplier: Ricca Chemical
RV010022-100N RV010022-500N
RCRV010022100NEA 142.2 CAD
RCRV010022100N RCRV010022500N
Germanium Standard, for AAS, VeriSpec®, RICCA Chemical Company
Standards Cationic Standards Germanium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml