Erbium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010379-500N RV010379-250N RV010379-100N
RCRV010379100NEA 408.64 CAD
RCRV010379100N RCRV010379250N RCRV010379500N
Erbium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Erbium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml