Copper Standard, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010377-100N
RV010377-250N
RV010377-500N
RCRV010377100NEA
409.22
CAD
RCRV010377100N
RCRV010377250N
RCRV010377500N
Copper Standard, for ICP, VeriSpec®, Ricca Chemical Company
Standards
Cationic Standards
Copper Standards
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml