Bismuth Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010268-100N RV010268-250N RV010268-500N RV010370-100N RV010370-500N
RCRV010268100NEA 188.02 CAD
RCRV010268100N RCRV010268250N RCRV010268500N RCRV010370100N RCRV010370500N
Bismuth Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Bismuth Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml