Bismuth Standard, for AAS, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010008-100N RV010008-500N
RCRV010008100NEA 110.6 CAD
RCRV010008100N RCRV010008500N
Bismuth Standard, for AAS, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Bismuth Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml