Antimony Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010310-100N RV010310-250N RV010310-500N RV010351-100N RV010351-250N RV010351-500N RV010412-100N RV010482-100N
RCRV010310100NEA 189.6 CAD
RCRV010310100N RCRV010310250N RCRV010310500N RCRV010351100N RCRV010351250N RCRV010351500N RCRV010412100N RCRV010482100N
Antimony Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Antimony Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml