Antimony Standards, for AAS, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010047-100N RV010047-500N RV010082-100N RV010082-500N
RCRV010047100NEA 110.6 CAD
RCRV010047100N RCRV010047500N RCRV010082100N RCRV010082500N
Antimony Standards, for AAS, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Antimony Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml