Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-F0141425 CS-F0141450 CS-F0141950 CS-F0141100 CS-F0149100 CS-F0144100 CS-F0141250 CS-F0149250 CS-F0144250 CS-F0144500
CA10201-810CS 97.43 CAD
CA10201-810 CA10201-812 CA10201-814 CA10201-816 CA10201-818 CA10201-820 CA10201-822 CA10201-824 CA10201-826 CA10201-828
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Flasks Round Bottom Flasks
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
  • An outer diameter between 5 and 7.5 mm
  • Heavy wall design
With a threaded side port for insertion of thermometer or bleed tubes.
Order Now
SPECIFICATIONS
Environmentally Preferable