ChemScience Filter Tubes with Fritted Disc, Glass Stopcock, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
CS-F0351420C CS-F0351420M CS-F0351430C CS-F0351430M CS-F0352440C CS-F0352440M
CA10201-520CS 435.06 CAD
CA10201-520 CA10201-522 CA10201-524 CA10201-526 CA10201-528 CA10201-530
ChemScience Filter Tubes with Fritted Disc, Glass Stopcock, SATI International Scientific
Filters Filter Tubes
With two 2 mm glass stopcocks having constant pressure retention devices.