CORIO™ CP Immersion Heating Circulators, JULABO USA, Inc.
Supplier: JULABO
9013000.22 9013000.33 9013000.22 9013000.33
76353-866EA 5280.63 CAD
76353-866 76353-868 CA76353-866 CA76353-868
CORIO™ CP Immersion Heating Circulators, JULABO USA, Inc.
Circulating Thermostats Open Circulator Systems Thermostat Controllers
CORIO™ CP immersion heating circulator with adjustable clamp for internal temperature applications to 50 L from 20 to 200 °C.
  • Infinitely adjustable pressure pump
  • Bright LED display
  • USB and RS232 ports
For internal temperature applications. Clamps easily on bath tanks with walls to 30 mm thick. Immersion depth: 7.5 to 16.5 cm.
CORIO CP circulator features a bright LED display button controls. The USB and RS232 ports facilitate external communication to PC or PLC systems. Pump connections: M16×1 male threads.
Certifications: CE certified.
Delivery information: Includes CORIO CP immersion circulator and adjustable clamp. Supplied without accessories.