PHCbi MPR Series Pharmaceutical Refrigerators with Freezer, PHC Corporation
Supplier: PHC Corporation
MPR-N450FH-PA MPR-N450FSH-PA MPR-N250FH-PA MPR-N250FSH-PA MPR-N250FH-PA MPR-N250FSH-PA MPR-N450FH-PA MPR-N450FSH-PA
76353-814EA 15805.85 CAD
76353-814 76353-816 76498-952 76498-954 CA76498-952 CA76498-954 CA76353-814 CA76353-816
PHCbi MPR Series Pharmaceutical Refrigerators with Freezer, PHC Corporation
Refrigerators
The PHCbi brand Pharmaceutical Refrigerators with Freezer offers a complete storage solution in one cabinet. These multipurpose cabinets are designed for safe, reliable storage of vaccines and biological materials.
  • Refrigerated and frozen products can be maintained within a single footprint
  • Optimum temperature uniformity under full load conditions
  • Independent refrigeration systems
  • The microprocessor controller and softkey interface are door mounted at eye level
  • Adjustable audible and visual alarms
  • Slim profile cabinet design
  • Data transfer is through a USB port
Hydrocarbon refrigerants help achieve facility sustainability objectives by minimizing any environmental impact without compromising cooling performance, ambient tolerance and recovery speeds following door openings.
The refrigerator includes three adjustable, tempered glass shelves. The solid shelf construction eliminates tipping of bottles and vials. A rear wall LED light is included for product viewing.
Both models include manual defrost for the freezer compartment. The refrigerator compartment includes an automatic defrost, which activates electronically when needed. The refrigerator evaporator operates above freezing at all times. This prevents vaccines and laboratory supplies from freezing.