Papanicolaou's solution modified Easypap, VWR® Stain_histology, cytology
Supplier: VWR International
|
Danger
|
Papanicolaou stains are used in conjunction with Hematoxylin nuclear stains in the diagnosis of malignant cytological diseases.
- Crisp, clear staining
- Well-defined cytoplasm
- ready-to-use
Gynecological staining protocols use OG-6 and hematoxylin along with either EA-36 or EA-50. Non gynecological procedures require use of EA-65. EASYpap is a single step cytology staining solution. Combination of EA and OG-6 solutions saves times by providing customer with an easy one step process. FDA approved for in vitro diagnostic use. For Professional Use Only.
Storage Temperature:
Ambient |
UN:
1170 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...