Papanicolaou's solution EA 50, VWR® Stain_histology, cytology

Supplier: VWR International
Danger

10143-158 10143-186
10143-158CS 6560.82 CAD
10143-158 10143-186
Papanicolaou's solution EA 50, VWR® Stain_histology, cytology
Papanicolaou's solution EA 50

Papanicolaou stains are used in conjunction with Hematoxylin nuclear stains in the diagnosis of malignant cytological diseases.


  • Crisp, clear staining
  • Well-defined cytoplasm
  • ready-to-use


Gynecological staining protocols use OG-6 and hematoxylin along with either EA-36 or EA-50. Non gynecological procedures require use of EA-65. EASYpap is a single step cytology staining solution. Combination of EA and OG-6 solutions saves times by providing customer with an easy one step process. FDA approved for in vitro diagnostic use. For Professional Use Only.

Storage Temperature: Ambient
MDL Number: MFCD00147831
UN: 1170
ADR: 3 II

Order Now

Specification Test Results

Absorbance (Visible Scan 700- 400nm) @626 nm 370 – 500 MA
Absorbance (Visible Scan 700- 400nm) @ 528nm 270 – 350 MA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR