Hematoxylin solution (Harris), VWR® stain for histology, for cytology mercury-free, acidified
Supplier: VWR International
|
Warning
|
10143-610
10143-612
10143-610CS
3318.7
CAD
10143-610
10143-612
Hematoxylin solution (Harris), VWR® stain for histology, for cytology mercury-free, acidified
Hematoxylin staining solutions
General purpose nuclear stain, regressive type. Used with hematoxylin and eosin staining.
- Crisp, clear staining
- Well-defined cytoplasm
- ready-to-use
FDA approved for in vitro diagnostic use. For Professional Use Only.
Formula:
C₁₆H₁₄O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 |
Specification Test Results
Absorbance (Visible Scan 700- 400nm) @~560 nm | 170-230 MA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...