Hematoxylin solution_SC (Harris), VWR® Stain_histology, cytology Hg_free

Supplier: VWR International
Warning

Synonyms: DcH-48, Natural Black I, Hydroxybrasilin

10143-606 10143-608
10143-606CS 6898.03 CAD
10143-606 10143-608
Hematoxylin solution_SC (Harris), VWR® Stain_histology, cytology Hg_free
Hematoxylin staining solutions

General purpose nuclear stain, regressive type. Used with hematoxylin and eosin staining.


  • Crisp, clear staining
  • Well-defined cytoplasm
  • ready-to-use


FDA approved for in vitro diagnostic use. For Professional Use Only.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Specification Test Results

Absorbance (Visible Scan 700- 400nm) @~560 nm ≥600 MA

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR