Hematoxylin solution Gill III, VWR® stain for histology, for cytology

Supplier: VWR International
Warning

Synonyms: HydroxybrasilinDcH-48Natural Black I

7016 7017
10143-152CS 6621.92 CAD
10143-152 10143-150
Hematoxylin solution Gill III, VWR® stain for histology, for cytology
Hematoxylin staining solutions

Gill 1X is ideal for routine cytology staining. This single strength formulation optimally stains gynecological and non-gynecological specimens. General purpose nuclear stain, progressive type. Used with hematoxylin and eosin staining. Gill 2X and Gill 3X should be used when a stronger or darker nuclear stain is required for histology samples.


  • Crisp, clear staining
  • Well-defined cytoplasm
  • ready-to-use


FDA approved for in vitro diagnostic use. For Professional Use Only.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Specification Test Results

Absorbance (Visible Scan 700- 400nm) @~560 nm ≥375 MA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR