Hareta Hydrophilic IHC Microscope Slides, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
IHC-A90BL IHC-A90PK IHC-A90WT IHC-A90BL-1B IHC-A90PK-1B IHC-A90WT-1B
76339-794CS 604.27 CAD
76339-794 76339-796 76339-798 76339-800 76339-802 76339-804
Hareta Hydrophilic IHC Microscope Slides, Springside Scientific
Slides Microscope Slides
Affordable, pure white glass slides specifically made for IHC applictions.
  • Strong electrostatically charged surface
  • Two years of testing in the Ventana staining system
  • Pure white, ultra-clear glass
Hydrophilic glass will withstand extreme staining environments.
Order Now
SPECIFICATIONS
Environmentally Preferable