Eosin Y (yellowish) 1% alcoholic solution, VWR® stain for histology, for cytology

Supplier: VWR International
Danger

Synonyms: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt

7006-G
10143-132CS 3026.33 CAD
10143-132
Eosin Y (yellowish) 1% alcoholic solution, VWR® stain for histology, for cytology
Eosin Y (yellowish)

Eosin Y 1% Alcoholic is a cytoplasmic stain that when used in the hematoxylin and eosin staining procedure provides excellent definition of cell components in shades of red and pink.


  • Crisp, clear staining
  • Well-defined cytoplasm
  • ready-to-use


Eosin Y 1% Alcoholic is a cytoplasmic stain that when used in the hematoxylin and eosin staining procedure provides excellent definition of cell components in shades of red and pink. It is an excellent cytoplasmic stain which provides great contrast to the purple nuclei stained with hematoxylin. Intensified Eosin and Eosin-Phloxine have been formulated to provide the user with increased definition of the cytoplasmic components. These innovative modified eosin counterstains are substituted for Eosin Y 1% Alcoholic in the staining procedure. Timing may be varied to achieve the desired staining intensity. FDA approved for in vitro diagnostic use. For Professional Use Only.

Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
UN: 1170
ADR: 3,II
IVDLogo

Order Now

Specification Test Results

Absorbance (Visible Scan 700- 400nm) 410 – 430MA @~531 nm
pH@ 25°C 6.85 – 7.05
Filtration 1 micron nominal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR