WypAll® Waterless Cleaning Wipes Refill, Kimberly-Clark Professional
Supplier: Kimberly-Clark
WYPALL®
91367 91367
75801-474CS 299.95 CAD
75801-474 CA75801-474
WypAll® Waterless Cleaning Wipes Refill, Kimberly-Clark Professional
Wipes Disposable Wipes and Towels Presaturated Wipes
WypAll® waterless cleaning wipes are the ideal pre-saturated cleaner for on-the-go cleaning of hands, tools, parts, and non-porous work surfaces.
  • Wipes are pre-saturated with an alcohol-free natural citrus cleaning agent
  • Tough on grease, but gentle enough to use on hands
  • Wipes contain moisturizers to help keep skin hydrated
  • One textured side for scrubbing dirt, grime, and grease (great for tool cleaning)
  • One smooth side for wiping dirt off surfaces and hands
The alcohol-free natural citrus solvents cut through grease, grime, and oil, while soothing ingredients moisturize the skin. The specially designed cleaning fabric incorporates a textured surface on one side to increase scrubbing action for more thorough cleaning (even a tool with grime caked on), while being thin and flexible enough to get into tight spaces. The other side features a smooth surface, which wipes away dirt from hands or work surfaces. The middle absorbent layer soaks up and holds the dirt.
These wipes are truly a must-have for any dirty job and cleaning on the go. When effectiveness and value top the list of criteria, trust WypAll® to deliver.
Packaging: Wipes are packaged in 6 refill packs of 75 sheets, with 450 sheets per case.