Automatic Shoe Cover Dispensers and Shoe Covers, Shoe Inn™
Supplier: Shoe Inn
SI-7150 7HBD-70HC 7FAB-80HC 7FWT-100HC 7FAB-100HC 7FWT-80HC SI-7200 7SUP-80HC 7PLA-110HC
76336-914CS 791.26 CAD
76336-914 76336-926 76336-912 76336-924 76336-910 76336-922 76336-920 76336-918 76336-916
Automatic Shoe Cover Dispensers and Shoe Covers, Shoe Inn™
PPE Dispensers
Automatic shoe cover dispensers make putting shoe covers on faster, easier, safer, and cleaner.
  • Increase efficiency and productivity
  • Reduce worker's compensation injuries
  • Favored by regulators and inspectors
  • Improve compliance
  • Avoid contamination
  • Ergonomically friendly
  • No electricity required
Automatic shoe cover dispensers were designed to provide a faster, easier, safer, and cleaner method of donning shoe covers. Customers will love the ease of use these revolutionary dispensers provide, as will any regulators and inspectors that visit the facilities utilizing these devices.
Capacities range from 110 to 220 shoe covers. The ergonomic automatic shoe cover dispensers do not require electricity to operate, allowing them to be placed anywhere. Additionally, they are very easy and quick to load and use.
Certifications: Approved for use in cleanrooms up to Class 100 and Class 5 per Federal Standard 209E and ISO STD. 14644-1, as certified by Ultraclean Products Approval Laboratory.
Ordering information: Shoe cover cases come with 15 cartridges.