Wright-Giemsa stain, VWR® Quick III™ Haematology

Supplier: VWR International

Synonyms: Wright-Giemsa Solution

10143-232 10143-238
10143-232CS 6734.61 CAD
10143-232 10143-238
Wright-Giemsa stain, VWR® Quick III™ Haematology
Wright-Giemsa stain (solutions & kits)

VWR® brand hematology stain offerings satify the needs of laboratory professionals following manual, rapid stain or automated protocols.


  • Stain offerings for multiple protocols
  • Individual stains, reagents and packs have been used in the automated market on the Hematek®, Midas®, Sysmex® and Sakura® instruments


Single-step stains are offered in Wright's and Wright Giemsa formulations and use deionized water to complete the staining process. Three step stain offering yields results comparable to Wright Giemsa stain with results available as soon as 15 seconds.


Certifications: Hematology stain products are manufactured using dyes certified by the Biological Stain Commission. All products undergo Quality Control evaluation using fresh blood.

Order Now

Specification Test Results

Quick III Solution II Absorbance 440-460MN@~650nm
Quick III Solution II pH 6.85-6.95@25°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR