Wright-Giemsa stain, VWR® for hematology

Supplier: VWR International

Synonyms: Wright-Giemsa Solution

5583 5584
10143-162CS 3918.51 CAD
10143-162 10143-160
Wright-Giemsa stain, VWR® for hematology
Wright-Giemsa stain (solutions & kits)

VWR® brand hematology stain offerings satify the needs of laboratory professionals following manual, rapid stain or automated protocols.


  • Stain offerings for multiple protocols
  • Individual stains, reagents and packs have been used in the automated market on the Hematek®, Midas®, Sysmex® and Sakura® instruments


Single-step stains are offered in Wright's and Wright Giemsa formulations and use deionized water to complete the staining process. Three step stain offering yields results comparable to Wright Giemsa stain with results available as soon as 15 seconds.


Certifications: Hematology stain products are manufactured using dyes certified by the Biological Stain Commission. All products undergo Quality Control evaluation using fresh blood.

Order Now

Specification Test Results

Absorbance Eosin Visible Scan-(700- 400nm) 325– 375 MA@525 nm
Absorbance MB/Azure Visible Scan (700-400nm) 580-680 MA @650 nm
Performance supersedes absorbance specifications and is the final criteria for acceptance.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR