Alcohol 95%, VWR®, Reagent Grade for histology
Supplier: VWR International
|
Danger
|
Synonyms:
Reagent alcohol
ES34114VWR
89370-082CS
162.01
CAD
89370-082
Alcohol 95%, VWR®, Reagent Grade for histology
Alcohol
Reagent Grade Alcohol, 95%
VWR Histology Reagents are manufactured from high quality raw materials in an ISO 13485 facility, ensuringoptimal performance in your histology lab.
Storage Temperature:
Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 UN: 1987 ADR: 3,II |
Specification Test Results
Components | |
Ethanol – 190 Proof | 89.5-91.5% |
Methanol | 4.0-5.0% |
Isopropanol | 4.5-5.5% |
Characteristics | |
Methanol and Ethyl Alcohol (SDA 3A 190 proof) (v/v) | 94.0-96.0% |
Isopropyl Alcohol (2-Propanol) (v/v) | 4.0-6.0% |
Specific Gravity @20°C | 0.810-0.812 |
Apparent Proof @20°C | Lot Analysis |
Color (Pt-Co) | 10 max. |
Acidity | To Pass |
Odor | Characteristic |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...