Dichloroacetic acid (DCA) 3% (v/v) in toluene, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

BR674-20 BR674-4X2.5GL
BJBR674-20EA 2393.7 CAD
BJBR674-20 CABJBR674425GL
Dichloroacetic acid (DCA) 3% (v/v) in toluene, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Dichloroacetic acid (DCA)

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination


UNSPSC Code: 12350000

Formula: C₂H₂Cl₂O₂
Density: 8,815 g/cm3
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR