Trichloroacetic acid (TCA) 3% w/v dichloromethane, BioSyn™ Detritylation Reagent, DNA/RNA synthesis, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: TCA

BR605-2 BR605-4 BR605-20 BR605-4X2.5L
CABJBR605-2CS 1749.79 CAD
CABJBR605-2 CABJBR605-4 BJBR605-20 CABJBR6054X2.5
Trichloroacetic acid (TCA) 3% w/v dichloromethane, BioSyn™ Detritylation Reagent, DNA/RNA synthesis, Burdick & Jackson™
Trichloroacetic acid (TCA)

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination


UNSPSC Code: 12350000

Formula: Cl₃CCOOH
MW: 163.39 g/mol
Boiling Pt: 196
Melting Pt: –95 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 113 Does not flash. Closed cup
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
UN: 2922
ADR: 8 II
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR