Capping Reagent 90/10 Tetrahydrofuran/N-Methylimidazole (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
BR651-2
BR651-4X2.5L
BR651-4
CABJBR651-2CS
4415.29
CAD
CABJBR651-2
CABJBR6514X2.5
CABJBR651-4
Capping Reagent 90/10 Tetrahydrofuran/N-Methylimidazole (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
UNSPSC Code: 12350000
Boiling Pt:
66 °C (main component) Melting Pt: –108.5 °C (main component) Density: 0.888 g/cm³ (20 °C) (main component) Flash Pt: –15 °C Storage Temperature: Ambient |
UN:
2924 ADR: 3,II |