Water with 0,1% (v/v) formic acid, LabReady™, Solvent Blend LC-MS, HPLC, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA

LC452-1 LC452-2.5
CABJLC452-1CS 657.17 CAD
CABJLC452-1 CABJLC452-2.5
Water with 0,1% (v/v) formic acid, LabReady™, Solvent Blend LC-MS, HPLC, Burdick & Jackson™
Water with 0,1% (v/v) formic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk


UNSPSC Code: 12350000

Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 64-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR