N,N-Dimethylformamide ≥99.8% (by GC) for peptide synthesis, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
NSC 5356
, Formic acid dimethylamide
, Dimethylformamide
, DMFA
, DMF
, Formyldimethylamine
, N-Formyldimethylamine
34440-2.5L
34440-2.5L
CABJ34440-2.5LEA
346.97
CAD
BJ34440-2.5L
CABJ34440-2.5L
N,N-Dimethylformamide ≥99.8% (by GC) for peptide synthesis, Burdick & Jackson™
N,N-Dimethylformamide
- Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
- As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
- Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
Beilstein Number: 605365
Formula:
HCON(CH₃)₂ MW: 73.09 g/mol Boiling Pt: 152…153 °C (1.013 hPa) Melting Pt: –61 °C Density: 0.94 g/cm³ (20 °C) Flash Pt: 57.5 °C (1.013.25 hPa) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III |
Specification Test Results
Assay (GC) | Min. 99.8 % |
Water (Karl Fischer) | Max. 0.02 % |
Non-volatile matter | Max. 0.001 % |
Amines (as dimethylamine) | Max. 0.0002 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...