Triethylamine ≥99.5%, BioSyn™ for HPLC, Fluka™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: (Diethylamino)ethane, TEA

17924-10X2ML 17924-1L
CABJ17924102MLCS 270.67 CAD
CABJ17924102ML CABJ17924-1L
Triethylamine ≥99.5%, BioSyn™ for HPLC, Fluka™
Triethylamine

Beilstein Number: 605283
RTECS Number: YE0175000
UNSPSC Code: 12352116

Formula: (C₂H₅)₃N
MW: 101.19 g/mol
Boiling Pt: 90
Melting Pt: -115
Density: 0.73 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3 II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) min. 99.5 %
Suitability test (HPLC) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR