DuoClick™ Specimen Containers with Temperature Strip, Parter Medical Products
Supplier: Parter Medical
143024 243024
76333-278CS 339.66 CAD
76333-278 76333-280
DuoClick™ Specimen Containers with Temperature Strip, Parter Medical Products
Sample Containers General Purpose Containers
The DuoClick™ system helps ensure an audible notification of correct closure. Specialty containers with temperature strip allows for temperature monitoring of specimen in drug screening and toxicology applications.
  • DuoClick™ lid alerts user of correct, leak-proof closure
  • Leak Resistant
  • Graduation markings for accurate volume measurement
Specimen containers are the ideal choice for collection and transport of specimens in microbiology, urinalysis, pathology/histology, and drug testing laboratories. All containers are available sterile or non-sterile, and with many lids, labeling, and packaging options. Simply turn the lid until you hear the two 'clicks' to fully ensure 95 kPa compliance.
Certifications: 95 kPa certified.