Hei-VAP Ultimate Control, Rotary Evaporators, Heidolph®
Supplier: Heidolph NA, LLC
036041060 036041061 036041062 036041063 036041064 036041065 036041066 036041067 036041068 036041069 036041070 036041071 036041072 036041073 036041074 036041075 036041076 036041077 036041078 036041079
76338-440EA 23504.08 CAD
76338-440 76338-442 76338-444 76338-446 76338-448 76338-450 76338-452 76338-454 76338-456 76338-458 76338-460 76338-462 76338-464 76338-466 76338-468 76338-470 76338-472 76338-474 76338-476 76338-478
Hei-VAP Ultimate Control, Rotary Evaporators, Heidolph®
Evaporators Rotary Evaporators
Designed as precise setting of rotation speed and heating bath temperature on the digital touch display or for quick access during an active process, directly via the knobs on the operating panel.
  • Clear display of all values
  • Central operating panel with 7" touch display
  • With timer, limitation of maximum temperature, multilingual menu and text display for error messages
  • LED ring lights and pictogram to display the activity even from a distance
  • USB and Micro SD plus LAN / RS 232 interfaces
  • Optional display of vapor temperature
It has integrated solvent library to facilitate adjustment of the required vacuum on an external vacuum controller with timer, limitation of maximum temperature, multilingual menu and text display for error messages. Available in Hand Lift (HL) and Motor Lift (ML) Configurations.
Ordering information: G1 glassware set is a diagonal coil condenser. G3 glassware set is a vertical coil condenser. G3 XL glassware is a higher evaporation rate condensor. Glassware set G5 is a dry ice condenser. Glassware set G6 is a vertical condenser.