Hei-VAP Expert Rotary Evaporators, Heidolph®
Supplier: Heidolph NA, LLC
036041000 036041001 036041002 036041003 036041004 036041005 036041006 036041007 036041008 036041009 036041010 036041011 036041012 036041013 036041014 036041015 036041016 036041017 036041018 036041019
76338-320EA 17030.5 CAD
76338-320 76338-322 76338-324 76338-326 76338-328 76338-330 76338-332 76338-334 76338-336 76338-338 76338-340 76338-342 76338-344 76338-346 76338-348 76338-350 76338-352 76338-354 76338-356 76338-358
Hei-VAP Expert Rotary Evaporators, Heidolph®
Evaporators Rotary Evaporators
Designed as precise setting of rotation speed and heating bath temperature on the digital touch display or for quick access during an active process, directly via the knobs on the operating panel.
  • Clear display of all values
  • Central operating panel with 5" touch display
  • Separate knobs for rotation and heating with locking function to prevent accidental readjusting
  • LED ring lights and pictogram to display the activity even from a distance
  • USB and Micro SD interfaces
  • Optional display of vapor temperature
  • Subsequent upgrade to Hei-VAP Expert Control possible,
More Product Information
Order Now
SPECIFICATIONS
Model Hei-VAP Expert, HL, G1 Hei-VAP Expert, HL, G3 Hei-VAP Expert, HL, G3 XL Hei-VAP Expert, HL, G5 Hei-VAP Expert, HL, G6 Hei-VAP Expert, HL, G1B Hei-VAP Expert, HL, G 3B Hei-VAP Expert, HL, G 3B XL Hei-VAP Expert, HL, G5 B Hei-VAP Expert, HL, G6 B Hei-VAP Expert, ML, G1 Hei-VAP Expert, ML, G3 Hei-VAP Expert, ML, G3 XL Hei-VAP Expert, ML, G5 Hei-VAP Expert, ML, G6 Hei-VAP Expert, ML, G1B Hei-VAP Expert, ML, G3 B Hei-VAP Expert, ML, G3 B XL Hei-VAP Expert, ML, G5 B Hei-VAP Expert, ML, G6 B
Environmentally Preferable
Rotation speed 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm 10 - 280 rpm
Heating capacity 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W 1300 W
Cooling surface 1400 cm² 1400 cm² 2200 cm² 1400 cm² 1400 cm² 1400 cm² 1400 cm² 2200 cm² 1400 cm² 1400 cm² 1400 cm² 1400 cm² 2200 cm² 1400 cm² 1400 cm² 1400 cm² 1400 cm² 2200 cm² 1400 cm² 1400 cm²
Bath capacity 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L 4.5 L
Protection class DIN EN 60529 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20 IP 20
Timer Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes
Temperature range 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C 20…210 °C
Max. flask capacity 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l 5 l
Bath Ø 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm 255 mm
Accuracy ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C ±1 °C