Statfree® Clear Dissipative Bags, Desco Industries
Supplier: Desco
13871 13873 13874 13881 13883 13884 13880
89131-020PK 18.71 CAD
89131-020 89131-024 89131-026 89131-028 89131-032 89131-034 89131-038
Statfree® Clear Dissipative Bags, Desco Industries
Bags Storage Bags
Statfree® Clear Dissipative Bags can be used for packaging ESD sensitive items or non-ESD items in an ESD protected area.
Clear dissipative polyester film bags for packaging ESD sensitive items or non-ESD items in ESD protected areas. Dissipative inner and outer surfaces: Rs 1×10⁴ to <1×10¹¹ Ω per ANSI/ESD STM11.11. Allows bag to be used throughout an ESD protected area without risking damage to ESD susceptible products. No Amines or N-octanoic Acid Compatible with polycarbonate contents. Printed with ESD protective symbol and lot code information.
Certifications: Meets the requirements of ANSI/ESD S541 for quality control purposes. Lead-free RoHS compliant.
Packaging: Statshield® bags are packaged 100 per package in an oversized shielding bag rather than a cardboard box. The bags are not exposed to water vapors that will degrade the metallized shielding layer.