Reztore® Antistatic Coating, Desco
Supplier: Desco Industries
Reztore™
10415
89131-018EA 42.25 CAD
89131-018
Reztore® Antistatic Coating, Desco
Sprays
Reztore® Antistatic Coating keeps triboelectric charges well below 200 volts per EIA 625.
  • Long lasting hard coat formula
  • Static dissipative
  • Non-toxic
  • Removes contaminating particulate
  • Rtt: 10E6 ohms to < 1×10E9 ohms per ANSI/ESD S4.1
  • Bag-in-box packaging available
  • Lead-free RoHS compliant
  • Made in The United States of America
Long lasting hard coat formula protection lasts three times as long as other leading brands. Non-toxic and safe to use coating provides a safe path to ground. Removes contaminating particulates and eliminates possible ESD event. This product has been certified to meet the RoHS requirement.