Adjustable Metal Wrist Strap, Desco
Supplier: Desco
91070 09041 09042
48888-323PK 50.48 CAD
48888-323 89129-452 89129-454
Adjustable Metal Wrist Strap, Desco
Grounders Wrist Grounders
Adjustable metal wrist straps are made of medical grade series stainless steel backplate and links.
  • Metal expansion wristband
  • Patented size adjustment
  • Medical Grade series stainless steel backplate and links
  • Solid stainless steel machined snap stud
  • Date coded
  • Made in United States of America
Metal Expansion Wristband used with a coil cord to create a path-to-ground for an operator with resistance <3.5×107 ohms per the required limits of ANSI/ESD S20.20, tested per ANSI/ESD S1.1 or ESD TR53. Adjustable design to allow the operator to adjust the size to their wrist. Medical grade series stainless steel backplate and links create a more reliable path-to-ground for the operator. Solid stainless steel machined snap stud provides a consistent 1 to 5 pound release, to prevent accidental disconnects between wristband and cord. Date coded so the lifespan of the product may be tracked and to ensure quality control.
Certifications: UL listed.