RACK’N’LAB Sample Bag Carriers, Labplas
Supplier: Labplas
SA5-7012 SA5-3070
75849-330CS 949.15 CAD
75849-330 75849-332
RACK’N’LAB Sample Bag Carriers, Labplas
Carriers Transport Carriers
The RACK’N’LAB is best used to transport bags used in food testing and environmental water sampling amongst many other industries. Its innovative design allows multiple bags to be used whilst being held upright without the need for bottles or specifically designed bags.
  • Made of FDA approved virgin polypropylene
  • Shipped in one inch thick boxes with three flat racks inside
  • Assembled or dismantled in less than one minute
  • The sturdy handle facilitates safe transportation
  • Identified rows allow for easy identification of samples
  • Up to 40 ml can be retained in
More Product Information