Capinsert™ Color Coding Inserts, Simport Scientific
Supplier: Simport Scientific
T345Y T345W T345R T345L T345B
CA73960-178CS 40 CAD
CA73960-178 CA73960-180 CA73960-182 CA73960-184 CA73960-186
Capinsert™ Color Coding Inserts, Simport Scientific
Cap Inserts
Capinserts™ can be used for all color coding needs.
  • Made of polypropylene
  • With write-on frosted area
  • Available in ten colors
The Capinsert™ is used to color code a multitude of Simport® products according to your specific needs. It is inserted on top of the closure and has a write-on frosted area for sample identification.