AlphaTec® 53-001 Series Gloves, Ansell
Supplier: Ansell Healthcare
53001070-NA 53001090-NA 53001100-NA 53001080-NA
76339-854BG 162.46 CAD
76339-854 76339-858 76339-860 76339-862 76339-874
AlphaTec® 53-001 Series Gloves, Ansell
Gloves
One glove with the benefits of two, offering multi-barrier chemical protection.
  • Innovative multi-layer polymer design: nitrile/neoprene/nitrile
  • Provides protection against a wide range of chemicals, from acids and bases to hydrocarbons and organic solvents
  • Tested according to EN ISO 374:2016; permeation time >30 minutes against 13 chemicals out of 18 listed
  • Features the latest MICROCHEM™ chemical barrier technology, providing superior protection for use in hazardous environments
  • ANSELL GRIP™ technology provides enhanced dexterity, grip, and comfort when
More Product Information