BRAND® Inserts 2 in 1 for Cell Culture, BrandTech® Scientific, Inc.

Supplier: Brandtech

782711 782716 782717 782700 782701 782706 782707 782720 782721 782726 782727 782730 782710 782731 782736 782737 782740 782741 782746 782747 782750 782751 782756 782757
76337-186PK 652.42 CAD
76337-186 76337-188 76337-190 76337-192 76337-194 76337-196 76337-198 76337-200 76337-202 76337-204 76337-206 76337-208 76337-224 76337-226 76337-228 76337-230 76337-232 76337-234 76337-236 76337-238 76337-240 76337-242 76337-244 76337-246
BRAND® Inserts 2 in 1 for Cell Culture, BrandTech® Scientific, Inc.
Microplates Insert Plates and Inserts
BRAND® insert 2 in 1 offers unlimited flexibility for 2D and 3D cell culture. Inserts can be used as a free standing or hanging insert for co-culture, air-lift cultures, migration assays and more.

  • cellGrade™ plus surface for enhanced cell attachment
  • PC or PET membrane
  • Available in 0.4 µm, 1 µm, 3 µm, or 8 µm pore sizes
  • Compatible with all common 6-, 12-, or 24-well plates and tissue culture dishes

Common applications include co-culture, trans-migration, chemotaxis, transport/barrier analysis (TEER), cytotoxicity and air-lift culture assays.

Pyrogen and endotoxin free according to LAL test, detection limit 0.01 EU/ml.

Certifications: Manufactured in a Class 8 cleanroom, in accordance with ISO 14644-1. Sterile according to ISO 11137, SAL 10-⁶. Non-cytoxic according to ISO 10993-5.

Packaging: Inserts are available individually packed or in multi-packs.

Caution: These products can expose you to chemicals including α-Methyl styrene (alpha-Methylstyrene), which is known to cause cancer, and Di-n-butyl Phthalate (DBP), which is known to cause birth defects or other reproductive harm.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR